Learn More
Medchemexpress LLC 5-methoxyresorcinol | 2174-64-3 | MFCD00002285 | 99.6% | 140.14 g/mol | C7H8O3 | 250 MG
SDP
Catalog No. 5000302487
Encompass_Preferred
5-Methoxyresorcinol is a phenolic compound provided as a laboratory standard and synthetic intermediate. Supplied as an off-white to yellow solid with a reported purity of 99.63%, it is suitable for analytical reference, method development, and use in organic synthesis.
  • Off-white to yellow solid physical form.
  • Reported purity of 99.63% suitable for analytical work.
  • Commonly used as an analytical standard and synthetic intermediate.
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Available in small pack sizes appropriate for bench-scale research.
Catalog No. 50-003-02487 Supplier Medchemexpress LLC Supplier No. 250MGHYW015752
5-Methoxyresorcinol is a phenolic compound provided as a laboratory standard and synthetic intermediate. Supplied as an off-white to yellow solid with a reported purity of 99.63%, it is suitable for analytical reference, method development, and use in organic synthesis.
  • Off-white to yellow solid physical form.
  • Reported purity of 99.63% suitable for analytical work.
  • Commonly used as an analytical standard and synthetic intermediate.
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Available in small pack sizes appropriate for bench-scale research.