Learn More
Medchemexpress LLC 5-methoxyresorcinol | 2174-64-3 | MFCD00002285 | 99.6% | 140.14 g·mol⁻¹ | C7H8O3 | 500 MG
SDP
Catalog No. 5000300072
Encompass_Preferred
5-Methoxyresorcinol (CAS 2174-64-3) is a research chemical and analytical standard used to model the resorcinol-type A ring of flavonoids for synthetic and mechanistic studies. It is a well-characterized solid (C7H8O3, MW 140.14 g·mol⁻1) commonly used as an analytical reference in laboratory workflows.
  • Used as a model molecule for flavonoid A-ring chemistry.
  • Serves as an analytical standard for LC- and GC-based methods.
  • Solid, well-characterized compound (C7H8O3; MW 140.14 g·mol⁻1).
  • High purity suitable for research and analytical applications.
  • Offered in multiple pack sizes for laboratory use.
Catalog No. 50-003-00072 Supplier Medchemexpress LLC Supplier No. HYW015752500MG
5-Methoxyresorcinol (CAS 2174-64-3) is a research chemical and analytical standard used to model the resorcinol-type A ring of flavonoids for synthetic and mechanistic studies. It is a well-characterized solid (C7H8O3, MW 140.14 g·mol⁻1) commonly used as an analytical reference in laboratory workflows.
  • Used as a model molecule for flavonoid A-ring chemistry.
  • Serves as an analytical standard for LC- and GC-based methods.
  • Solid, well-characterized compound (C7H8O3; MW 140.14 g·mol⁻1).
  • High purity suitable for research and analytical applications.
  • Offered in multiple pack sizes for laboratory use.