Learn More
Medchemexpress LLC 5-Methoxysalicylic acid (5-MeOSA) | 2612-02-4 | MFCD00002459 | 168.15 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00785610MM/1ML
Encompass_Preferred
5-Methoxysalicylic acid (5-MeOSA) is a natural compound used as a useful matrix in the MALDI MS analysis of oligonucleotides when combined with spermine. It finds application in various biochemical studies and analytical procedures.
  • Natural compound
  • Useful matrix for MALDI MS analysis
  • Can be combined with spermine for oligonucleotide analysis
  • Applicable in biochemical studies
  • Suitable for analytical procedures
Catalog No. 50-004-46811