Learn More
Apexbio Technology LLC 5-Methoxytryptamine 608-07-1 100mg
SDP
Catalog No. 502240718
Encompass_Preferred
5-Methoxytryptamine (CAS 608-07-1) also known as Mexamine or Methoxytryptamine is a synthetic derivative of tryptamine It acts as a full agonist at multiple serotonin receptor subtypes including 5-HT1 5-HT2 5-HT4 5-HT6 and 5-HT7 Through these interactions 5-Methoxytryptamine modulates serotonergic signaling pathways making it a valuable tool for studying serotonin-mediated physiological and neuropharmacological processes This compound is commonly utilized in research exploring serotonergic receptor function neural signaling and associated behavioral or biochemical outcomes
This item is not returnable. View return policy
Catalog No. 50-224-0718 Supplier Apexbio Technology LLC Supplier No. B7872100
5-Methoxytryptamine (CAS 608-07-1) also known as Mexamine or Methoxytryptamine is a synthetic derivative of tryptamine It acts as a full agonist at multiple serotonin receptor subtypes including 5-HT1 5-HT2 5-HT4 5-HT6 and 5-HT7 Through these interactions 5-Methoxytryptamine modulates serotonergic signaling pathways making it a valuable tool for studying serotonin-mediated physiological and neuropharmacological processes This compound is commonly utilized in research exploring serotonergic receptor function neural signaling and associated behavioral or biochemical outcomes