Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Methoxytryptamine

Catalog No. 286583100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-658-3100MG 100 mg
28-658-31G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 28-658-3100MG Supplier Sigma Aldrich Supplier No. 286583100MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

5-Methoxytryptamine

Specifications

CAS 608-07-1
Melting Point 121°C to 123°C (lit.)
Molecular Formula C11H14N2O
Linear Formula C11H14N2O
MDL Number MFCD00005662
Quantity 100 mg
Synonym 2-(5-Methoxyindol-3-yl)ethylamine; 3-(2-Aminoethyl)-5-methoxyindole; 5-MOT; 5-Methoxyindole-3-ethanamine; Deacetylmelatonin; NSC 56422
Molecular Weight (g/mol) 190.24
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.