Learn More
Apexbio Technology LLC 5-Methoxytryptamine 608-07-1 500mg
SDP
Catalog No. 5000090048
Encompass_Preferred
5-Methoxytryptamine (CAS 608-07-1) also known as Mexamine or Methoxytryptamine is a synthetic derivative of tryptamine It acts as a full agonist at multiple serotonin receptor subtypes including 5-HT1 5-HT2 5-HT4 5-HT6 and 5-HT7 Through these interactions 5-Methoxytryptamine modulates serotonergic signaling pathways making it a valuable tool for studying serotonin-mediated physiological and neuropharmacological processes This compound is commonly utilized in research exploring serotonergic receptor function neural signaling and associated behavioral or biochemical outcomes
Catalog No. 50-000-90048 Supplier Apexbio Technology LLC Supplier No. B7872500
5-Methoxytryptamine (CAS 608-07-1) also known as Mexamine or Methoxytryptamine is a synthetic derivative of tryptamine It acts as a full agonist at multiple serotonin receptor subtypes including 5-HT1 5-HT2 5-HT4 5-HT6 and 5-HT7 Through these interactions 5-Methoxytryptamine modulates serotonergic signaling pathways making it a valuable tool for studying serotonin-mediated physiological and neuropharmacological processes This compound is commonly utilized in research exploring serotonergic receptor function neural signaling and associated behavioral or biochemical outcomes