Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-methoxytryptamine hydrochloride | 66-83-1 | MFCD00012684 | 99.6% | 226.70 | C11H15ClN2O | 1 ML
SDP

Catalog No. 5000302501
Encompass_Preferred

5-Methoxytryptamine hydrochloride is the hydrochloride salt of 5-methoxytryptamine (O-methylserotonin), a nonselective 5-HT receptor agonist and endogenous metabolite of melatonin. Supplied as a ready-to-use solution in DMSO, it is suitable for in vitro receptor pharmacology and serotonergic signaling studies.

  • Ready-to-use 10 mM solution in DMSO for in vitro assays.
  • Nonselective 5-HT receptor agonist, useful for serotonergic studies.
  • High chemical purity (reported ~99.6%).
  • Molecular weight 226.70; formula C11H15ClN2O.
  • Storage guidance: 4°C sealed; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-003-02501 Supplier Medchemexpress LLC Supplier No. HYW015169A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Methoxytryptamine hydrochloride is the hydrochloride salt of 5-methoxytryptamine (O-methylserotonin), a nonselective 5-HT receptor agonist and endogenous metabolite of melatonin. Supplied as a ready-to-use solution in DMSO, it is suitable for in vitro receptor pharmacology and serotonergic signaling studies.

  • Ready-to-use 10 mM solution in DMSO for in vitro assays.
  • Nonselective 5-HT receptor agonist, useful for serotonergic studies.
  • High chemical purity (reported ~99.6%).
  • Molecular weight 226.70; formula C11H15ClN2O.
  • Storage guidance: 4°C sealed; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.