Learn More
Medchemexpress LLC 5-methoxytryptamine hydrochloride | 66-83-1 | MFCD00012684 | 99.6% | 226.70 g/mol | C11H15ClN2O | 250 MG
SDP
Catalog No. 5000302443
Encompass_Preferred
5-Methoxytryptamine hydrochloride is the hydrochloride salt of 5-methoxytryptamine, a metabolite of melatonin that functions as a nonselective 5-HT (serotonin) receptor agonist. It is provided as a research-grade reagent for studies of serotonin receptor activity, antioxidant effects, and radioprotective actions in biochemical and neuroscience applications.
  • Provides a research reagent for serotonin receptor assays.
  • Acts as a nonselective 5-HT receptor agonist.
  • Exhibits antioxidant and radioprotective properties.
  • Formulated as a hydrochloride salt to aid solubility in common solvents.
  • Stable under recommended storage: 4°C (solid); in solvent, -80°C (6 months) or -20°C (1 month).
  • Characterized by molecular weight 226.70 g/mol and formula C11H15ClN2O.
Catalog No. 50-003-02443 Supplier Medchemexpress LLC Supplier No. 250MGHYW015169A
5-Methoxytryptamine hydrochloride is the hydrochloride salt of 5-methoxytryptamine, a metabolite of melatonin that functions as a nonselective 5-HT (serotonin) receptor agonist. It is provided as a research-grade reagent for studies of serotonin receptor activity, antioxidant effects, and radioprotective actions in biochemical and neuroscience applications.
  • Provides a research reagent for serotonin receptor assays.
  • Acts as a nonselective 5-HT receptor agonist.
  • Exhibits antioxidant and radioprotective properties.
  • Formulated as a hydrochloride salt to aid solubility in common solvents.
  • Stable under recommended storage: 4°C (solid); in solvent, -80°C (6 months) or -20°C (1 month).
  • Characterized by molecular weight 226.70 g/mol and formula C11H15ClN2O.