Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-methoxytryptamine hydrochloride | 66-83-1 | MFCD00012684 | 99.6% | 226.70 g/mol | C11H15ClN2O | 500 MG
SDP

Catalog No. 5000303161
Encompass_Preferred

5-Methoxytryptamine hydrochloride is the hydrochloride salt of 5-methoxytryptamine, a melatonin metabolite and nonselective serotonin (5-HT) receptor agonist supplied for laboratory research. It is provided as a solid for in vitro and in vivo studies and intended for research use only.

  • Hydrochloride salt form for improved stability and solubility in aqueous media.
  • Well characterized with molecular weight 226.70 g/mol and formula C11H15ClN2O.
  • Soluble in DMSO, PEG300, Tween-80, and saline for flexible formulation options.
  • Storage recommendations: sealed, moisture-free at 4°C; in solution store at -20°C to -80°C depending on duration.
  • High purity reported on supplier COA (≈99.6%), suitable for research applications.
  • Used in studies of serotonin signaling and melatonin metabolism.

Catalog No. 50-003-03161 Supplier Medchemexpress LLC Supplier No. HYW015169A500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Methoxytryptamine hydrochloride is the hydrochloride salt of 5-methoxytryptamine, a melatonin metabolite and nonselective serotonin (5-HT) receptor agonist supplied for laboratory research. It is provided as a solid for in vitro and in vivo studies and intended for research use only.

  • Hydrochloride salt form for improved stability and solubility in aqueous media.
  • Well characterized with molecular weight 226.70 g/mol and formula C11H15ClN2O.
  • Soluble in DMSO, PEG300, Tween-80, and saline for flexible formulation options.
  • Storage recommendations: sealed, moisture-free at 4°C; in solution store at -20°C to -80°C depending on duration.
  • High purity reported on supplier COA (≈99.6%), suitable for research applications.
  • Used in studies of serotonin signaling and melatonin metabolism.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.