Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methoxytryptamine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog No. AAL1907006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1907006 5 g
AAL1907003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1907006 Supplier Thermo Scientific Chemicals Supplier No. L1907006
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-83-1
Molecular Formula C11H15ClN2O
Molecular Weight (g/mol) 226.704
MDL Number MFCD00012684
InChI Key TXVAYRSEKRMEIF-UHFFFAOYSA-N
Synonym 5-methoxytryptamine hydrochloride, mexamine hydrochloride, o-methylserotonin hydrochloride, 2-5-methoxy-1h-indol-3-yl ethanamine hydrochloride, 3-2-aminoethyl-5-methoxyindole hydrochloride, unii-4265ib6qek, 5-methoxy-1h-indole-3-ethylamine hydrochloride, 1h-indole-3-ethanamine, 5-methoxy-, monohydrochloride, 5-methoxy tryptamine hcl, 5 methoxytryptamine hydrochloride
PubChem CID 6198
IUPAC Name 2-(5-methoxy-1H-indol-3-yl)ethanamine;hydrochloride
SMILES COC1=CC2=C(C=C1)NC=C2CCN.Cl

Specifications

Melting Point 245°C to 250°C (decomposition)
Odor Odorless
Quantity 5 g
Beilstein 3717598
Sensitivity Hygroscopic
Merck Index 14,6002
Formula Weight 226.71
Percent Purity ≥98%
Chemical Name or Material 5-Methoxytryptamine hydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 200-637-6
RTECSNumber NL4110000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.