Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Methoxytryptophol | 712-09-4 | 99.95% | 191.23 | 100 MG
SDP

Catalog No. 5000436363
Encompass_Preferred

5-Methoxytryptophol is a natural indole present in the pineal gland. It is for research use only and not sold to patients.

  • Human endogenous metabolite
  • Purity of 99.95%
  • Molecular weight of 191.23
  • Molecular formula C11H13NO2
  • CAS number 712-09-4
  • Appears as a viscous liquid
  • Colorless to light yellow
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-004-36363 Supplier Medchemexpress LLC Supplier No. HY113440100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Methoxytryptophol is a natural indole present in the pineal gland. It is for research use only and not sold to patients.

  • Human endogenous metabolite
  • Purity of 99.95%
  • Molecular weight of 191.23
  • Molecular formula C11H13NO2
  • CAS number 712-09-4
  • Appears as a viscous liquid
  • Colorless to light yellow
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.