Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-methyl-1,3-thiazole-2-carbaldehyde

Catalog No. CDS011813 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS011813 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS011813 Supplier Sigma Aldrich Supplier No. CDS011813100MG

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-heptanol is the main biotransformation product of n-heptane.3-heptanol can be used:
  • As a solvent to form microenvironments around single-walled carbon nanotubes
  • To prepare substituted pyrimidine derivatives as c1 domain-targeted isophthalate analogs to study their binding affinities towards pkcα
  • Isoform
  • As a building block to synthesize 4-(3-adamantan-1-yl-ureido)-butyric acid and cyclohexanecarboxylic acid derivatives as seh inhibitors

Specifications

Molecular Formula C5H5NOS
Linear Formula C5H5NOS
MDL Number MFCD08060733
Quantity 25 mL
Molecular Weight (g/mol) 127.16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.