Learn More
Medchemexpress LLC 5-(methyl-13C)pyrimidine-2,4(1H,3H)-dione-1,3-15N2 | 99.9% | 129.09 | C413CH615N2O2 | 5mg
SDP
Catalog No. 5000268286
Encompass_Preferred
Thymine-15N2 13C is the 13C and 15N labeled Thymine (HY-W010450) Thymine is one of the four nucleobases in the nucleic acid of DNA and can be a target for actions of 5-Fluorouracil (HY-90006) in cancer treatment with a Km of 2 3 M[1]
Catalog No. 50-002-68286 Supplier Medchemexpress LLC Supplier No. HYW7472145MG
Thymine-15N2 13C is the 13C and 15N labeled Thymine (HY-W010450) Thymine is one of the four nucleobases in the nucleic acid of DNA and can be a target for actions of 5-Fluorouracil (HY-90006) in cancer treatment with a Km of 2 3 M[1]