Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic acid | 956317-36-5 | 100.0% | C10H9N3O2 | 25 G
SDP

Catalog No. 5000358460
Encompass_Preferred

5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic acid is a chemical compound supplied as a white to off-white solid with a purity of 99.99%. It is stable under recommended storage conditions and is intended for use as a laboratory chemical and in the manufacture of substances.

  • High purity
  • White to off-white solid form
  • Melting point between 175.4-175.8°C
  • Boiling point of 413°C at 760 mmHg
  • Flash point of 204°C
  • Chemically stable under recommended storage conditions
  • Identified for laboratory chemical use
  • Identified for the manufacture of substances

Catalog No. 50-003-58460 Supplier Medchemexpress LLC Supplier No. 25GHY77001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic acid is a chemical compound supplied as a white to off-white solid with a purity of 99.99%. It is stable under recommended storage conditions and is intended for use as a laboratory chemical and in the manufacture of substances.

  • High purity
  • White to off-white solid form
  • Melting point between 175.4-175.8°C
  • Boiling point of 413°C at 760 mmHg
  • Flash point of 204°C
  • Chemically stable under recommended storage conditions
  • Identified for laboratory chemical use
  • Identified for the manufacture of substances

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.