Learn More
Medchemexpress LLC 5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic acid | 956317-36-5 | 99.99% | C10H9N3O2 | 10 G
SDP
Catalog No. 5000433732
Encompass_Preferred
5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic acid is a chemical compound used as a laboratory reagent and in the manufacture of other substances. This white to off-white solid is characterized by its high purity and specific storage requirements.
  • High purity (99.99%)
  • Suitable for laboratory chemicals and manufacture of other substances
  • Stable as powder at -20°C for 3 years or 4°C for 2 years
  • In solvent, stable at -80°C for 6 months or -20°C for 1 month
  • Appears as a white to off-white solid
Catalog No. 50-004-33732 Supplier Medchemexpress LLC Supplier No. HY7700110G
5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic acid is a chemical compound used as a laboratory reagent and in the manufacture of other substances. This white to off-white solid is characterized by its high purity and specific storage requirements.
  • High purity (99.99%)
  • Suitable for laboratory chemicals and manufacture of other substances
  • Stable as powder at -20°C for 3 years or 4°C for 2 years
  • In solvent, stable at -80°C for 6 months or -20°C for 1 month
  • Appears as a white to off-white solid