Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic acid | 956317-36-5 | MFCD18382679 | >99.99% | C10H9N3O2 | 500 G
SDP

Catalog No. 5000434078
Encompass_Preferred

5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic acid is a solid chemical intended for laboratory use and substance manufacturing. It has a melting point of 175.4-175.8°C, a boiling point of 413°C at 760 mmHg, and a flash point of 204°C. The product is stable under recommended storage conditions.

  • Intended for laboratory chemicals
  • Suitable for manufacture of substances
  • Solid appearance
  • Melting point: 175.4-175.8°C
  • Boiling point: 413°C at 760 mmHg
  • Flash point: 204°C
  • Stable under recommended storage conditions

Catalog No. 50-004-34078 Supplier Medchemexpress LLC Supplier No. HY77001500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic acid is a solid chemical intended for laboratory use and substance manufacturing. It has a melting point of 175.4-175.8°C, a boiling point of 413°C at 760 mmHg, and a flash point of 204°C. The product is stable under recommended storage conditions.

  • Intended for laboratory chemicals
  • Suitable for manufacture of substances
  • Solid appearance
  • Melting point: 175.4-175.8°C
  • Boiling point: 413°C at 760 mmHg
  • Flash point: 204°C
  • Stable under recommended storage conditions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.