Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic acid | 956317-36-5 | MFCD18382679 | C10H9N3O2 | 100 G
SDP

Catalog No. 5000433734
Encompass_Preferred

5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic acid is a chemical compound primarily used as an intermediate in organic synthesis and medicinal chemistry, particularly in the synthesis of Suvorexant. It is intended for laboratory research purposes.

  • Used as an intermediate in organic synthesis.
  • Utilized in medicinal chemistry.
  • Key component in the synthesis of Suvorexant.
  • Stable under recommended storage conditions.
  • Available as a solid.
  • Melting point: 174-176 °C.
  • Boiling point: 413 °C.

Catalog No. 50-004-33734 Supplier Medchemexpress LLC Supplier No. HY77001100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic acid is a chemical compound primarily used as an intermediate in organic synthesis and medicinal chemistry, particularly in the synthesis of Suvorexant. It is intended for laboratory research purposes.

  • Used as an intermediate in organic synthesis.
  • Utilized in medicinal chemistry.
  • Key component in the synthesis of Suvorexant.
  • Stable under recommended storage conditions.
  • Available as a solid.
  • Melting point: 174-176 °C.
  • Boiling point: 413 °C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.