Learn More
Medchemexpress LLC HY-W012078 100mg Medchemexpress, 5-Methyl-2'-deoxycytidine CAS:838-07-3 Purity:>98%
SDP
Catalog No. 501934558
Encompass_Preferred
Medchemexpress, HY-W012078 100mg 5-Methyl-2'-deoxycytidine CAS:838-07-3 5-Methyl-2'-deoxycytidine in single-stranded DNA can act in cis to signal de novo DNA methylation [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4558 Supplier Medchemexpress LLC Supplier No. HYW012078100MG
Medchemexpress, HY-W012078 100mg 5-Methyl-2'-deoxycytidine CAS:838-07-3 5-Methyl-2'-deoxycytidine in single-stranded DNA can act in cis to signal de novo DNA methylation [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.