Learn More
Medchemexpress LLC 5-methyl-2'-deoxypseudouridine | 65358-15-8 | 99.4% | 242.23 | C10H14N2O5 | 1 MG
SDP
Catalog No. 5000247991
Encompass_Preferred
Pseudothymidine is a C-nucleoside analog of thymidine supplied as a powdered research reagent. It serves as an alternative nucleoside substrate for molecular biology and nucleic acid studies and includes analytical data and recommended storage conditions for laboratory use.
  • C-nucleoside analog of thymidine.
  • Molecular formula C10H14N2O5 and molecular weight 242.23.
  • High purity: 99.4%.
  • Available in small milligram package sizes (1 mg, 5 mg, 10 mg).
  • Powder form with storage guidance: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months) or -20°C (up to 1 month).
  • For research use only; not for clinical or patient use.
Catalog No. 50-002-47991 Supplier Medchemexpress LLC Supplier No. HY1019691MG
Pseudothymidine is a C-nucleoside analog of thymidine supplied as a powdered research reagent. It serves as an alternative nucleoside substrate for molecular biology and nucleic acid studies and includes analytical data and recommended storage conditions for laboratory use.
  • C-nucleoside analog of thymidine.
  • Molecular formula C10H14N2O5 and molecular weight 242.23.
  • High purity: 99.4%.
  • Available in small milligram package sizes (1 mg, 5 mg, 10 mg).
  • Powder form with storage guidance: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months) or -20°C (up to 1 month).
  • For research use only; not for clinical or patient use.