Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-methyl-2-furanmethanol | 3857-25-8 | MFCD00130134 | 112.13 g/mol | C6H8O2 | 1 ML
SDP

Catalog No. 5000208315
Encompass_Preferred

5-Methyl-2-furanmethanol is a naturally occurring furan derivative used as a research standard and analytical reference. It is typically supplied as a concentrated stock solution for laboratory use and can be diluted to working concentrations.

  • Natural product used as a reference standard for research and analysis.
  • Molecular formula C6H8O2 and molecular weight 112.13 g/mol.
  • Soluble in DMSO; stock solutions commonly prepared at 1 mM, 5 mM, or 10 mM.
  • In vivo vehicle solubility reported ≥2.5 mg/mL (≈22.30 mM) in several vehicles.
  • Storage: solid at -20°C for long-term stability; in solvent store at -80°C for extended storage.
  • Available in small pack sizes suitable for standards and analytical use.

Catalog No. 50-002-08315 Supplier Medchemexpress LLC Supplier No. HYW08791910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Methyl-2-furanmethanol is a naturally occurring furan derivative used as a research standard and analytical reference. It is typically supplied as a concentrated stock solution for laboratory use and can be diluted to working concentrations.

  • Natural product used as a reference standard for research and analysis.
  • Molecular formula C6H8O2 and molecular weight 112.13 g/mol.
  • Soluble in DMSO; stock solutions commonly prepared at 1 mM, 5 mM, or 10 mM.
  • In vivo vehicle solubility reported ≥2.5 mg/mL (≈22.30 mM) in several vehicles.
  • Storage: solid at -20°C for long-term stability; in solvent store at -80°C for extended storage.
  • Available in small pack sizes suitable for standards and analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.