Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Methyl-2'-O,4'-C-methylenecytidine | 847650-87-7 | 98.0% | 269.25 | C11H15N3O5 | 1 ML
SDP

Catalog No. 5000281061
Encompass_Preferred

5-Methyl-2'-O,4'-C-methylenecytidine is a cytidine nucleoside analog supplied for research use, provided as a solid and as a ready-to-use 10 mM solution in DMSO for studies of DNA methylation and epigenetic mechanisms.

  • Cytidine nucleoside analog for DNA methylation and epigenetics research.
  • High purity (98.0%) suitable for analytical applications.
  • Available as solid and as a 10 mM solution in DMSO for convenience.
  • Molecular weight 269.25 for accurate dosing and calculations.
  • Recommended storage: solid at 4°C; in solvent at -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical use.

Catalog No. 50-002-81061 Supplier Medchemexpress LLC Supplier No. HYW41129510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Methyl-2'-O,4'-C-methylenecytidine is a cytidine nucleoside analog supplied for research use, provided as a solid and as a ready-to-use 10 mM solution in DMSO for studies of DNA methylation and epigenetic mechanisms.

  • Cytidine nucleoside analog for DNA methylation and epigenetics research.
  • High purity (98.0%) suitable for analytical applications.
  • Available as solid and as a 10 mM solution in DMSO for convenience.
  • Molecular weight 269.25 for accurate dosing and calculations.
  • Recommended storage: solid at 4°C; in solvent at -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.