Learn More
Medchemexpress LLC 5-Methyl-2-O-methyl 10mM 1mL
SDP
Catalog No. 5000204810
Encompass_Preferred
5-Methyl-2 -O-methyl-uridine (2 -O-methyl-5-methyluridine) is a thymidine analog Analogs of this series have insertional activity towards replicated DNA They can be used to label cells and track DNA synthesis 5-Methyl-2 -O-methyl-uridine is a modified nucleoside that can be found in tRNA isolated from rabbit liver[1][2]
Catalog No. 50-002-04810 Supplier Medchemexpress LLC Supplier No. HYW10023410MM/1ML
5-Methyl-2 -O-methyl-uridine (2 -O-methyl-5-methyluridine) is a thymidine analog Analogs of this series have insertional activity towards replicated DNA They can be used to label cells and track DNA synthesis 5-Methyl-2 -O-methyl-uridine is a modified nucleoside that can be found in tRNA isolated from rabbit liver[1][2]