Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-methyl-2-phenyl-2H-1,2,3-triazole-4-carbonyl chloride, Thermo Scientific™

Catalog No. CC16202DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC16202DE 5 g Amber Glass Bottle
CC16202CB 250 mg Amber Glass Bottle
CC16202DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC16202DE Supplier Thermo Scientific Chemicals Supplier No. CC16202DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 36401-55-5
Molecular Formula C10H8ClN3O
Molecular Weight (g/mol) 221.64
MDL Number MFCD00052545
InChI Key UJYBUZMRRLFXGM-UHFFFAOYSA-N
Synonym 5-methyl-2-phenyl-2h-1,2,3-triazole-4-carbonyl chloride, 5-methyl-2-phenyl-1,2,3-triazole-4-carbonyl chloride, 2h-1,2,3-triazole-4-carbonylchloride, 5-methyl-2-phenyl, 5-methyl-2-phenyl-2h-1,2,3 triazole-4-carbonyl chloride, 5-methyl-2-phenyl-4-triazolecarbonyl chloride, 5-methyl-2-phenyl-2h-1,2,3 triazole-4-carbo nyl chloride, 5-methyl-2-phenyl-2h-1,2,3-triazole-4-carbonylchloride
PubChem CID 2776447
IUPAC Name 5-methyl-2-phenyltriazole-4-carbonyl chloride
SMILES CC1=NN(N=C1C(Cl)=O)C1=CC=CC=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 221.65
Percent Purity 97%
Chemical Name or Material 5-Methyl-2-phenyl-2H-1, 2, 3-triazole-4-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.