Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-2-phenylpyridine 98.0+%, TCI America™
SDP

Catalog No. M2969200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2969200MG 200 mg
M29691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2969200MG Supplier TCI America Supplier No. M2969200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27012-22-2
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.227
MDL Number MFCD04114180
InChI Key ZYLPQYYLLRBVOK-UHFFFAOYSA-N
Synonym 6-Phenyl-3-picoline
PubChem CID 47467
IUPAC Name 5-methyl-2-phenylpyridine
SMILES CC1=CN=C(C=C1)C2=CC=CC=C2

Specifications

Melting Point 55°C
Color White-Yellow
Boiling Point 166°C
Quantity 200 mg
Formula Weight 169.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl-2-phenylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.