Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-methyl-2-(pyrimidin-2-yl)benzoic acid | 1088994-22-2 | MFCD14706695 | 214.22 | 50 G
SDP

Catalog No. 5000447505
Encompass_Preferred

5-methyl-2-(pyrimidin-2-yl)benzoic acid is a chemical intermediate primarily utilized in the synthesis of anticancer drugs. This compound is typically found as a white to off-white solid.

  • White to off-white solid appearance
  • Molecular formula C12H10N2O2
  • Molecular weight 214.22
  • Boiling point 316 °C
  • Density 1.260
  • Flash point 145 °C
  • Heavy atoms count: 16
  • Rotatable bond count: 2
  • Number of rings: 2
  • Polar surface area: 63 Å2
  • Hydrogen bond acceptors count: 4
  • Hydrogen bond donors count: 1

Catalog No. 50-004-47505 Supplier Medchemexpress LLC Supplier No. HYW00387050G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-methyl-2-(pyrimidin-2-yl)benzoic acid is a chemical intermediate primarily utilized in the synthesis of anticancer drugs. This compound is typically found as a white to off-white solid.

  • White to off-white solid appearance
  • Molecular formula C12H10N2O2
  • Molecular weight 214.22
  • Boiling point 316 °C
  • Density 1.260
  • Flash point 145 °C
  • Heavy atoms count: 16
  • Rotatable bond count: 2
  • Number of rings: 2
  • Polar surface area: 63 Å2
  • Hydrogen bond acceptors count: 4
  • Hydrogen bond donors count: 1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.