Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-2-thiopheneboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. M18505G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1850-5G 5 g
M18501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1850-5G Supplier TCI America Supplier No. M18505G
Add to Cart
Add to Cart

Specifications

CAS 162607-20-7
Melting Point 150°C
Color Yellow
Molecular Formula C5H7BO2S
Quantity 5 g
MDL Number MFCD01318166
Synonym 5-methylthiophene-2-boronic acid, 5-methylthiophen-2-yl boronic acid, 5-methyl-2-thiopheneboronic acid, 5-methyl-2-thienyl boronic acid, 5-methyl-2-thienylboric acid
InChI Key NRIYPIBRPGAWDD-UHFFFAOYSA-N
SMILES B(C1=CC=C(S1)C)(O)O
IUPAC Name (5-methylthiophen-2-yl)boronic acid
Molecular Weight (g/mol) 141.979
PubChem CID 2734374
Formula Weight 141.98
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.