Learn More
Medchemexpress LLC 5-Methyl-2-thiophenecarboxaldehyde | 13679-70-4 | 99.9% | 126.18 | 100 G
SDP
Catalog No. 5000404483
Encompass_Preferred
5-Methyl-2-thiophenecarboxaldehyde acts as a candidate to microscopic third order non-linear optical (NLO) material. It is also an analytical standard intended for research and analytical applications.
  • Acts as a candidate to microscopic third order non-linear optical (NLO) material.
  • Analytical standard for research and analytical applications.
Catalog No. 50-004-04483 Supplier Medchemexpress LLC Supplier No. HY34465100G
5-Methyl-2-thiophenecarboxaldehyde acts as a candidate to microscopic third order non-linear optical (NLO) material. It is also an analytical standard intended for research and analytical applications.
  • Acts as a candidate to microscopic third order non-linear optical (NLO) material.
  • Analytical standard for research and analytical applications.