Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Methyl-2-thiophenecarboxaldehyde | 13679-70-4 | 99.9% | 126.18 | 25 G
SDP

Catalog No. 5000402809
Encompass_Preferred

5-Methyl-2-thiophenecarboxaldehyde is a chemical compound acting as a candidate for microscopic third-order non-linear optical (NLO) material. This compound is typically found as a yellow to brown liquid and is designated for research purposes only.

  • Candidate for microscopic third-order non-linear optical (NLO) material
  • High purity: 99.87%
  • Molecular weight: 126.18
  • Chemical formula: C6H6OS
  • Appearance: liquid
  • Color: yellow to brown
  • Originates from plants, Gramineae, Hordeum vulgare L.
  • Stable in pure form for up to 3 years at -20°C or 2 years at 4°C
  • Stable in solvent for up to 6 months at -80°C or 1 month at -20°C
  • Soluble in DMSO at 50 mg/mL
  • Intended for research use only

Catalog No. 50-004-02809 Supplier Medchemexpress LLC Supplier No. 25GHY34465
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Methyl-2-thiophenecarboxaldehyde is a chemical compound acting as a candidate for microscopic third-order non-linear optical (NLO) material. This compound is typically found as a yellow to brown liquid and is designated for research purposes only.

  • Candidate for microscopic third-order non-linear optical (NLO) material
  • High purity: 99.87%
  • Molecular weight: 126.18
  • Chemical formula: C6H6OS
  • Appearance: liquid
  • Color: yellow to brown
  • Originates from plants, Gramineae, Hordeum vulgare L.
  • Stable in pure form for up to 3 years at -20°C or 2 years at 4°C
  • Stable in solvent for up to 6 months at -80°C or 1 month at -20°C
  • Soluble in DMSO at 50 mg/mL
  • Intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.