Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-2-thiouracil 98.0+%, TCI America™
SDP

Catalog No. M099425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M099425G 25 g
M099410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M099425G Supplier TCI America Supplier No. M099425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 636-26-0
Molecular Formula C5H6N2OS
Molecular Weight (g/mol) 142.176
MDL Number MFCD00009777
InChI Key ZLAQATDNGLKIEV-UHFFFAOYSA-N
Synonym 4-Hydroxy-2-mercapto-5-methylpyrimidine, 4-Hydroxy-5-methyl-2-thiopyrimidine
PubChem CID 720471
IUPAC Name 5-methyl-2-sulfanylidene-1H-pyrimidin-4-one
SMILES CC1=CNC(=S)NC1=O

Specifications

Color White
Quantity 25 g
Formula Weight 142.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl-2-thiouracil
RTECSNumber XP2108500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.