Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-2-(trifluoromethyl)-3-furansulfonyl chloride, 97%, Thermo Scientific™

Catalog No. CC00703EA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC00703EA 10 g Amber Glass Bottle
CC00703DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC00703EA Supplier Thermo Scientific Chemicals Supplier No. CC00703EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 306935-02-4
Molecular Formula C6H4ClF3O3S
Molecular Weight (g/mol) 248.6
InChI Key GSYXWDYWBRICSD-UHFFFAOYSA-N
Synonym 5-methyl-2-trifluoromethyl furan-3-sulfonyl chloride, 5-methyl-2-trifluoromethyl-3-furansulfonyl chloride, 3-furansulfonylchloride, 5-methyl-2-trifluoromethyl, 5-methyl-2-trifluoromethyl furan-3-sulphonyl chloride, 5-methyl-2-trifluoromethyl-3-furansulfonylchloride, 5-methyl-2-trifluoromethyl-furan-3-sulfonyl chloride, chloro 5-methyl-2-trifluoromethyl 3-furyl sulfone, 3-furansulfonyl chloride, 5-methyl-2-trifluoromethyl-9ci
PubChem CID 2779895
IUPAC Name 5-methyl-2-(trifluoromethyl)furan-3-sulfonyl chloride
SMILES CC1=CC(=C(O1)C(F)(F)F)S(=O)(=O)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 248.61
Percent Purity 97%
Chemical Name or Material 5-Methyl-2-(trifluoromethyl)-3-furansulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.