Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-3-hexen-2-one (contains 5-Methyl-4-hexen-2-one), TCI America™
SDP

Catalog No. M236625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M236625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M236625ML Supplier TCI America Supplier No. M236625ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5166-53-0
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00012166
InChI Key IYMKNYVCXUEFJE-SNAWJCMRSA-N
PubChem CID 5363140
IUPAC Name (E)-5-methylhex-3-en-2-one
SMILES CC(C)C=CC(=O)C

Specifications

Color Yellow
Boiling Point 157°C
Quantity 25 mL
UN Number 1992
Formula Weight 112.17
Physical Form Liquid
Chemical Name or Material 5-Methyl-3-hexen-2-one (contains 5-Methyl-4-hexen-2-one)
RTECSNumber MP8750000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.