Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(5-Methyl-3-isoxazolyl)methylamine, 97+%, Thermo Scientific™

Catalog No. CC10413CB
Change view
Click to view available options
Quantity:
250 mg
5 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC10413CB 250 mg Amber Glass Bottle
CC10413DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC10413CB Supplier Thermo Scientific Chemicals Supplier No. CC10413CB
Add to Cart

Chemical Identifiers

CAS 154016-48-5
Molecular Formula C5H8N2O
Molecular Weight (g/mol) 112.132
InChI Key AZVWIMLQRLKLHH-UHFFFAOYSA-N
PubChem CID 2776306
IUPAC Name (5-methyl-1,2-oxazol-3-yl)methanamine
SMILES CC1=CC(=NO1)CN

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 5-methyl-3-isoxazolyl methylamine, 5-methylisoxazol-3-yl methanamine, 5-methyl-1,2-oxazol-3-yl methanamine, 5-methylisoxazol-3-yl methylamine, 1-5-methyl-1,2-oxazol-3-yl methanamine, 1-5-methylisoxazol-3-yl methanamine, 3-isoxazolemethanamine,5-methyl, 3-isoxazolemethanamine, 5-methyl, c-5-methyl-isoxazol-3-yl-methylamine, acmc-20a4az
Formula Weight 112.13
Percent Purity 97%
Chemical Name or Material (5-Methyl-3-isoxazolyl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.