Learn More
Medchemexpress LLC 5-Methyl-7-methoxyisoflavone | 82517-12-2 | 266.29 | 50 MG
SDP
Catalog No. 5000416028
Encompass_Preferred
5-Methyl-7-methoxyisoflavone is an orally active anti-oxidant with remyelinating activity. It inhibits the enzyme aromatase and acts as a non-steroidal anabolic isoflavone, demonstrating better potency in increasing muscle mass and endurance than Ipriflavone. It is also an inhibitor for NF-κB and can be used for fat loss, maintaining low cholesterol levels, and strengthening bones.
  • Orally active anti-oxidant
  • Inhibits aromatase enzyme
  • Non-steroidal anabolic isoflavone
  • Increases muscle mass and endurance
  • Supports fat loss
  • Helps maintain low cholesterol levels
  • Strengthens bones
  • Inhibits NF-κB
Catalog No. 50-004-16028 Supplier Medchemexpress LLC Supplier No. HYN199350MG
5-Methyl-7-methoxyisoflavone is an orally active anti-oxidant with remyelinating activity. It inhibits the enzyme aromatase and acts as a non-steroidal anabolic isoflavone, demonstrating better potency in increasing muscle mass and endurance than Ipriflavone. It is also an inhibitor for NF-κB and can be used for fat loss, maintaining low cholesterol levels, and strengthening bones.
  • Orally active anti-oxidant
  • Inhibits aromatase enzyme
  • Non-steroidal anabolic isoflavone
  • Increases muscle mass and endurance
  • Supports fat loss
  • Helps maintain low cholesterol levels
  • Strengthens bones
  • Inhibits NF-κB