Learn More
Medchemexpress LLC 5-methyl-7-methoxyisoflavone | 82517-12-2 | MFCD03093036 | 99.1% | 266.29 | C17H14O3 | 10 MG
SDP
Catalog No. 5000295936
Encompass_Preferred
5-methyl-7-methoxyisoflavone is a non-steroidal isoflavone used in research for its aromatase inhibitory and antioxidant properties; it has reported remyelinating activity and NF-κB inhibitory effects and is supplied in solid and DMSO solution formats for biochemical and cellular studies.
  • isoflavone scaffold with aromatase inhibitory activity.
  • Orally active antioxidant with reported remyelinating properties.
  • Shown to inhibit NF-κB signaling pathways.
  • High listed purity (approximately 99.1%).
  • Available as solid (10 mg-500 mg) and 10 mM DMSO solution formats for experimental flexibility.
Catalog No. 50-002-95936 Supplier Medchemexpress LLC Supplier No. HYN199310MG
5-methyl-7-methoxyisoflavone is a non-steroidal isoflavone used in research for its aromatase inhibitory and antioxidant properties; it has reported remyelinating activity and NF-κB inhibitory effects and is supplied in solid and DMSO solution formats for biochemical and cellular studies.
  • isoflavone scaffold with aromatase inhibitory activity.
  • Orally active antioxidant with reported remyelinating properties.
  • Shown to inhibit NF-κB signaling pathways.
  • High listed purity (approximately 99.1%).
  • Available as solid (10 mg-500 mg) and 10 mM DMSO solution formats for experimental flexibility.