Learn More
Cayman Chemical 5-methyl Chrysen 10mg
SDP
Catalog No. 502979397
Encompass_Preferred
A carcinogenic aryl hydrocarbon receptor agonist produced by the incomplete combustion of organic matter and found primarily in gasoline exhaust and tobacco smoke
Catalog No. 50-297-9397 Supplier Cayman Chemical Supplier No. 1867710 MG
A carcinogenic aryl hydrocarbon receptor agonist produced by the incomplete combustion of organic matter and found primarily in gasoline exhaust and tobacco smoke