Learn More
Zymo Research Corporation 5-Methyl dCTP (10mM)
SDP
Catalog No. 502037019
Encompass
Highlights:- High purity 5-Methyl dCTP- Readily incorporated into PCR amplicons with ZymoTaq™ or other DNA polymerases.- Free of endo-, exodeoxyribonuclease, ribonuclease,phosphatase and nicking activitiesApplications:PCR amplicons containing 5-methylcytosine are useful in a variety of applications such as:- The study of 5-methylcytosine binding/interacting proteins- Define methods that distinguish 5-methylcytosineConcentration: 1 µmol/100 µl as a [10 mM] solution.Storage Condition: 5-Methyl dCTP is guaranteed for 12 months at -20°C. Avoid multiple freeze/thaw cycles. For long-term storage, aliquoting is recommended with storage at -80°C.
Catalog No. 50-203-7019 Supplier Zymo Research Corporation Supplier No. D1035
Highlights:- High purity 5-Methyl dCTP- Readily incorporated into PCR amplicons with ZymoTaq™ or other DNA polymerases.- Free of endo-, exodeoxyribonuclease, ribonuclease,phosphatase and nicking activitiesApplications:PCR amplicons containing 5-methylcytosine are useful in a variety of applications such as:- The study of 5-methylcytosine binding/interacting proteins- Define methods that distinguish 5-methylcytosineConcentration: 1 µmol/100 µl as a [10 mM] solution.Storage Condition: 5-Methyl dCTP is guaranteed for 12 months at -20°C. Avoid multiple freeze/thaw cycles. For long-term storage, aliquoting is recommended with storage at -80°C.