Learn More
Chemgenes Corporation 5-Methyl-2'-deoxycytidine N, 100 g, Crystalline solid, ≥ 98% purity
SDP
Catalog No. NC2733352
Encompass
5-Methyl-2'-deoxycytidine (5mdC) is a crucial modified nucleoside that plays a significant role in epigenetic regulation and nucleic acid synthesis. It acts as a substrate for DNA methyltransferases, enhancing oligonucleotide stability and sensitivity. This compound is instrumental in various biological applications, particularly in therapeutic oligonucleotide development, where it helps manage immune responses and control gene expression by influencing DNA methylation patterns and promoter regulation.
  • Commonly used in epigenetics research
  • Improves stability of oligonucleotide duplexes
  • Enhances sensitivity of qPCR primers
  • Critical for DNA/RNA synthesis
  • Helps mitigate immune responses in therapeutic applications
  • Crystalline solid form with high purity
Catalog No. NC2733352 Supplier Chemgenes Corporation Supplier No. ANP6421100G
5-Methyl-2'-deoxycytidine (5mdC) is a crucial modified nucleoside that plays a significant role in epigenetic regulation and nucleic acid synthesis. It acts as a substrate for DNA methyltransferases, enhancing oligonucleotide stability and sensitivity. This compound is instrumental in various biological applications, particularly in therapeutic oligonucleotide development, where it helps manage immune responses and control gene expression by influencing DNA methylation patterns and promoter regulation.
  • Commonly used in epigenetics research
  • Improves stability of oligonucleotide duplexes
  • Enhances sensitivity of qPCR primers
  • Critical for DNA/RNA synthesis
  • Helps mitigate immune responses in therapeutic applications
  • Crystalline solid form with high purity