Learn More
Medchemexpress LLC 5-(methylamino)-2-naphthalen-1-yl-1,3-oxazole-4-carbonitrile | 847163-28-4 | MFCD04183120 | 99.2% | 249.27 g·mol⁻¹ | C15H11N3O | 5 MG
SDP
Catalog No. 5000263264
Encompass_Preferred
ML351 is a potent, selective small-molecule inhibitor of 15-lipoxygenase-1 (15-LOX-1) intended for laboratory research. It has been applied in cellular and preclinical in vivo models to probe 15-LOX-1-mediated lipid metabolism and to reduce β-cell oxidative stress. For research use only.
  • Potent inhibitor of 15-LOX-1 with reported IC50 of 200 nM.
  • High selectivity versus related isozymes (>250-fold), minimizing off-target activity.
  • High purity (≈99.2%) suitable for biochemical and cellular assays.
  • Soluble in common organic solvents and available in solid and solution formats for flexibility.
  • Suitable for in vitro and in vivo research on lipid metabolism and oxidative stress pathways.
Catalog No. 50-002-63264 Supplier Medchemexpress LLC Supplier No. HY1113105MG
ML351 is a potent, selective small-molecule inhibitor of 15-lipoxygenase-1 (15-LOX-1) intended for laboratory research. It has been applied in cellular and preclinical in vivo models to probe 15-LOX-1-mediated lipid metabolism and to reduce β-cell oxidative stress. For research use only.
  • Potent inhibitor of 15-LOX-1 with reported IC50 of 200 nM.
  • High selectivity versus related isozymes (>250-fold), minimizing off-target activity.
  • High purity (≈99.2%) suitable for biochemical and cellular assays.
  • Soluble in common organic solvents and available in solid and solution formats for flexibility.
  • Suitable for in vitro and in vivo research on lipid metabolism and oxidative stress pathways.