Learn More
Cayman Chemical 5-Methylcytidine, Purity: ≥98%, CAS: 2140-61-6, 100 mg
SDP
Catalog No. 502971168
Encompass_Preferred
A modified nucleoside derived from 5-methylcytosine and is a minor constituent of RNA as well as DNA for certain organisms; used in epigenetics research, especially in studies involving DNA methylation processes
Catalog No. 50-297-1168 Supplier Cayman Chemical Supplier No. 16111100 MG
A modified nucleoside derived from 5-methylcytosine and is a minor constituent of RNA as well as DNA for certain organisms; used in epigenetics research, especially in studies involving DNA methylation processes