Learn More
Selleck Chemical LLC 5-Methylcytidine
SDP
Catalog No. 5000004222
Encompass_Preferred
5-Methylcytidine is a modified nucleoside derived from 5-methylcytosine It is found in ribonucleic acids of animal plant and bacterial origin
Catalog No. 50-000-04222 Supplier Selleck Chemical LLC Supplier No. S61385MG
5-Methylcytidine is a modified nucleoside derived from 5-methylcytosine It is found in ribonucleic acids of animal plant and bacterial origin