Learn More
Medchemexpress LLC 5-Methylcytidine | 2140-61-6 | 99.9% | 257.24 | 1 ML
SDP
Catalog No. 5000436145
Encompass_Preferred
5-Methylcytidine is a nucleoside compound with antiviral activity, showing an IC50 against HSV-1 of 0.06 μM. It is also a common modification form in DNA, playing a role in gene expression regulation and development, and is associated with diseases like cancer.
  • Nucleoside compound
  • Antiviral activity against HSV-1
  • Common DNA modification
  • Involved in gene expression regulation and development
  • Related to disease occurrence and development, such as cancer
  • Potential for research in antiviral therapies
  • Useful for studying gene expression regulation and DNA modifications
  • Relevant for cancer research
Catalog No. 50-004-36145 Supplier Medchemexpress LLC Supplier No. HY11313510MM/1ML
5-Methylcytidine is a nucleoside compound with antiviral activity, showing an IC50 against HSV-1 of 0.06 μM. It is also a common modification form in DNA, playing a role in gene expression regulation and development, and is associated with diseases like cancer.
  • Nucleoside compound
  • Antiviral activity against HSV-1
  • Common DNA modification
  • Involved in gene expression regulation and development
  • Related to disease occurrence and development, such as cancer
  • Potential for research in antiviral therapies
  • Useful for studying gene expression regulation and DNA modifications
  • Relevant for cancer research