Learn More
Medchemexpress LLC 5-Methylcytidine | 2140-61-6 | 99.9% | 257.24 | 25 MG
SDP
Catalog No. 5000439081
Encompass_Preferred
5-Methylcytidine is a nucleoside compound with antiviral activity, exhibiting an IC50 of 0.06 μM against HSV-1. It is formed by the attachment of a methyl group to the 5th carbon atom of cytosine. 5-Methylcytidine is a common modification found in DNA and plays a significant role in processes such as gene expression regulation and development. It is also closely associated with the occurrence and development of diseases like cancer.
  • Nucleoside compound
  • Antiviral activity (IC50 against HSV-1 is 0.06 μM)
  • Plays a role in gene expression regulation and development
  • Related to diseases such as cancer
Catalog No. 50-004-39081 Supplier Medchemexpress LLC Supplier No. 25MGHY113135R
5-Methylcytidine is a nucleoside compound with antiviral activity, exhibiting an IC50 of 0.06 μM against HSV-1. It is formed by the attachment of a methyl group to the 5th carbon atom of cytosine. 5-Methylcytidine is a common modification found in DNA and plays a significant role in processes such as gene expression regulation and development. It is also closely associated with the occurrence and development of diseases like cancer.
  • Nucleoside compound
  • Antiviral activity (IC50 against HSV-1 is 0.06 μM)
  • Plays a role in gene expression regulation and development
  • Related to diseases such as cancer