Learn More
Medchemexpress LLC 5-Methylcytidine 5′-triphosphate trisodium | 327174-86-7 | 99.6% | 563.13 | 28 MG
SDP
Catalog No. 5000359986
Encompass_Preferred
5-Methylcytidine 5′-triphosphate (5-Methyl-CTP) trisodium is a modified nucleoside triphosphate. It can be used to replace unmodified mRNA, which leads to an increase in translational properties and stability, and a reduction in innate immune responses in human and other mammalian cells.
  • Increases translational properties
  • Enhances stability of mRNA
  • Reduces innate immune responses in mammalian cells
  • For research use only
Catalog No. 50-003-59986 Supplier Medchemexpress LLC Supplier No. HY147338A28MG
5-Methylcytidine 5′-triphosphate (5-Methyl-CTP) trisodium is a modified nucleoside triphosphate. It can be used to replace unmodified mRNA, which leads to an increase in translational properties and stability, and a reduction in innate immune responses in human and other mammalian cells.
  • Increases translational properties
  • Enhances stability of mRNA
  • Reduces innate immune responses in mammalian cells
  • For research use only