Learn More
Medchemexpress LLC 5-Methylcytidine 5′-triphosphate trisodium | 327174-86-7 | >99.6% | 563.13 | 57 MG
SDP
Catalog No. 5000359640
Encompass_Preferred
5-Methylcytidine 5′-triphosphate (5-Methyl-CTP) trisodium is a modified nucleoside triphosphate. It can be used in replacing unmodified mRNA, which leads to an increase in translational properties and stability, and a reduction in innate immune responses in human and other mammalian cells.
  • Modified nucleoside triphosphate
  • Increases translational properties and stability
  • Reduces innate immune responses in human and mammalian cells
Catalog No. 50-003-59640 Supplier Medchemexpress LLC Supplier No. HY147338A57MG
5-Methylcytidine 5′-triphosphate (5-Methyl-CTP) trisodium is a modified nucleoside triphosphate. It can be used in replacing unmodified mRNA, which leads to an increase in translational properties and stability, and a reduction in innate immune responses in human and other mammalian cells.
  • Modified nucleoside triphosphate
  • Increases translational properties and stability
  • Reduces innate immune responses in human and mammalian cells