Learn More
Cayman Chemical 5-Methylcytosine, Purity: ≥98%, CAS: 554-01-8, 10 g
SDP
Catalog No. 502915998
Encompass_Preferred
A methylated derivative cytosine; assoiated with reduced gene transcription; increased levels have been found found in non-expressed and untranslated DNA sequences; levels are increased on developmental genes to reduce aberrant expression in adults; found in the tRNA, mRNA, and rRNA from bacteria, archaea, and eukaryotes
Catalog No. 50-291-5998 Supplier Cayman Chemical Supplier No. 3874110 G
A methylated derivative cytosine; assoiated with reduced gene transcription; increased levels have been found found in non-expressed and untranslated DNA sequences; levels are increased on developmental genes to reduce aberrant expression in adults; found in the tRNA, mRNA, and rRNA from bacteria, archaea, and eukaryotes