Learn More
Cayman Chemical 5-MethylcytosIn 50g
SDP
Catalog No. 502917034
Encompass_Preferred
A phthalide with diverse biological activities increases levels of HO-1 in HepG2 cells as well as reduces hydrogen peroxide-induced increases in ROS and MDA in the same cells at 150 and 200 Ug/ml inhibits the proliferation of primary mouse aorta smooth muscle cells (IC50 0.1 Ug/ml) decreases bone resorption in ovariectomized mice at 15 mg/kg
Catalog No. 50-291-7034 Supplier Cayman Chemical Supplier No. 3874150 G
A phthalide with diverse biological activities increases levels of HO-1 in HepG2 cells as well as reduces hydrogen peroxide-induced increases in ROS and MDA in the same cells at 150 and 200 Ug/ml inhibits the proliferation of primary mouse aorta smooth muscle cells (IC50 0.1 Ug/ml) decreases bone resorption in ovariectomized mice at 15 mg/kg