Learn More
Apexbio Technology LLC 5-Methylcytosine 554-01-8 200mg
SDP
Catalog No. 5000090224
Encompass_Preferred
5-Methylcytosine (CAS 554-01-8) is a methylated derivative of cytosine commonly found as an epigenetic modification in both DNA and RNA across eukaryotic and prokaryotic organisms This modification regulates gene expression by influencing chromatin architecture and transcriptional activity thereby impacting cellular processes such as differentiation development and responses to environmental cues In plant systems such as Arabidopsis 5-Methylcytosine in RNA has been shown to modulate gene regulatory networks Due to its role in epigenetic regulation 5-Methylcytosine is widely utilized in studies investigating mechanisms of gene expression control and developmental biology
Catalog No. 50-000-90224 Supplier Apexbio Technology LLC Supplier No. C7031200
5-Methylcytosine (CAS 554-01-8) is a methylated derivative of cytosine commonly found as an epigenetic modification in both DNA and RNA across eukaryotic and prokaryotic organisms This modification regulates gene expression by influencing chromatin architecture and transcriptional activity thereby impacting cellular processes such as differentiation development and responses to environmental cues In plant systems such as Arabidopsis 5-Methylcytosine in RNA has been shown to modulate gene regulatory networks Due to its role in epigenetic regulation 5-Methylcytosine is widely utilized in studies investigating mechanisms of gene expression control and developmental biology