Learn More
Selleck Chemical LLC 5-Methylcytosine
SDP
Catalog No. 5000004234
Encompass_Preferred
5-Methylcytosine a methylated form of the DNA base cytosine is a major epigenetic mark in the nuclear DNA in mammals and may be involved in the regulation of gene transcription
Catalog No. 50-000-04234 Supplier Selleck Chemical LLC Supplier No. S602125MG
5-Methylcytosine a methylated form of the DNA base cytosine is a major epigenetic mark in the nuclear DNA in mammals and may be involved in the regulation of gene transcription