Learn More
Medchemexpress LLC 5-Methylcytosine-d4 | 1219795-15-9 | 129.15 | 5 MG
SDP
Catalog No. 5000448892
Encompass_Preferred
5-Methylcytosine-d4 is the deuterium labeled version of 5-Methylcytosine. 5-Methylcytosine is a known DNA modification in prokaryotes and eukaryotes. It forms symmetrical methylation on CpG dinucleotides in DNA, stabilizes tRNA/rRNA structure in RNA, and influences mRNA translation. It can be oxidized to produce 5hmC, 5fC, and 5caC. 5-Methylcytosine is used in epigenetics, developmental biology, and research on diseases such as colorectal cancer and hepatocellular carcinoma.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-48892 Supplier Medchemexpress LLC Supplier No. HYW008091S5MG
5-Methylcytosine-d4 is the deuterium labeled version of 5-Methylcytosine. 5-Methylcytosine is a known DNA modification in prokaryotes and eukaryotes. It forms symmetrical methylation on CpG dinucleotides in DNA, stabilizes tRNA/rRNA structure in RNA, and influences mRNA translation. It can be oxidized to produce 5hmC, 5fC, and 5caC. 5-Methylcytosine is used in epigenetics, developmental biology, and research on diseases such as colorectal cancer and hepatocellular carcinoma.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS