Learn More
Zymo Research Corporation 5-Methylcytosine dNTP Mix (10mM)
SDP
Catalog No. 50444100
Encompass
Applications: PCR amplicons containing 5-methylcytosine are useful in a variety of applications such as: - The study of 5-methylcytosine binding/interacting proteins - Define methods that detect 5-methylcytosine Concentration: 2.5 µmol/250 µl as a [10 mM] solution. Contains [2.5 mM] of each dNTP (dATP, dTTP, dGTP and d5mCTP). Storage Condition: 5-Methylcytosine dNTP Mix is guaranteed for 12 months at -20°C. Avoid multiple freeze/thaw cycles. For long-term storage, aliquoting is recommended with storage at -80°C.
Catalog No. 50-444-100 Supplier Zymo Research Corporation Supplier No. D1030
Applications: PCR amplicons containing 5-methylcytosine are useful in a variety of applications such as: - The study of 5-methylcytosine binding/interacting proteins - Define methods that detect 5-methylcytosine Concentration: 2.5 µmol/250 µl as a [10 mM] solution. Contains [2.5 mM] of each dNTP (dATP, dTTP, dGTP and d5mCTP). Storage Condition: 5-Methylcytosine dNTP Mix is guaranteed for 12 months at -20°C. Avoid multiple freeze/thaw cycles. For long-term storage, aliquoting is recommended with storage at -80°C.